2-Bromo-2,3,3,3-tetrafluoropropionamide 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC0131-1G
PC0131-5G
APOSPC0131-1GEA
6450
CZK
APOSPC0131-1G
APOSPC0131-5G
2-Bromo-2,3,3,3-tetrafluoropropionamide 97%
2-Bromo-2,3,3,3-tetrafluoropropionamide
Vzorec:
C₃H₂BrF₄NO MW: 223,952 g/mol Bod tání: 90…90,5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153719 CAS číslo: 422-22-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...