Tisk…

4-Amino-3,5-diiodopyridine

Dodavatel: Apollo Scientific
Varování

OR9959-1G OR9959-5G OR9959-10G
APOSOR9959-1GEA 662 CZK
APOSOR9959-1G APOSOR9959-5G APOSOR9959-10G
4-Amino-3,5-diiodopyridine
4-Amino-3,5-diiodopyridine
Vzorec: C₅H₄I₂N₂
MW: 345,908 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02663851
CAS číslo: 98136-86-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR