Tisk…

3,4-Dimethoxy-2-hydroxymethylpyridine

Dodavatel: Apollo Scientific
Varování

Synonyms: 3,4-Dimethylpicolinyl alcohol, (3,4-Dimethoxypyridin-2-yl)methanol

OR9947-25G OR9947-5G OR9947-100G
APOSOR9947-25GEA 2360 CZK
APOSOR9947-25G APOSOR9947-5G APOSOR9947-100G
3,4-Dimethoxy-2-hydroxymethylpyridine
3,4-Dimethoxy-2-hydroxymethylpyridine
Vzorec: C₈H₁₁NO₃
MW: 169.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06795941
CAS číslo: 72830-08-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR