1-[3-(Morpholin-4-yl)prop-1-yl]homopiperazine 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-[3-(1,4-Diazepan-1-yl)prop-1-yl]morpholine, 1-[3-(Morpholin-4-yl)prop-1-yl]-1,4-diazepane
OR9921-1G
OR9921-2G
OR9921-5G
APOSOR9921-1GEA
4010
CZK
APOSOR9921-1G
APOSOR9921-2G
APOSOR9921-5G
1-[3-(Morpholin-4-yl)prop-1-yl]homopiperazine 97%
1-[3-(Morpholin-4-yl)prop-1-yl]homopiperazine
Vzorec:
C₁₂H₂₅N₃O MW: 227,346 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03094710 CAS číslo: 827614-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...