L-(+)-Cystathionine
Dodavatel: Apollo Scientific
|
Varování
|
OR9895T-10MG
OR9895T-50MG
APOSOR9895T-10MGEA
6220
CZK
APOSOR9895T-10MG
APOSOR9895T-50MG
L-(+)-Cystathionine
L-(+)-Cystathionine
Intermediate in trans-sulfuration whereby the sulfur of methionine is transferred via homocysteine to cysteine. Collection Czechoslov.
Vzorec:
C₇H₁₄N₂O₄S Bod tání: 312 °C Storage Temperature: Freezer |
MDL Number:
MFCD00036685 CAS číslo: 56-88-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...