1-Methyl-4-(3-nitrobenzyl)piperazine 97%
Dodavatel: Apollo Scientific
OR9787-10G
OR9787-1G
OR9787-5G
APOSOR9787-10GEA
9860
CZK
APOSOR9787-10G
APOSOR9787-1G
APOSOR9787-5G
1-Methyl-4-(3-nitrobenzyl)piperazine 97%
1-Methyl-4-(3-nitrobenzyl)piperazine
Vzorec:
C₁₂H₁₇N₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD01466619 CAS číslo: 198281-54-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...