Tisk…

2-(Hydroxymethyl)-5-phenoxyfuran 97%

Dodavatel: Apollo Scientific

Synonyms: (Phenoxy-2-furyl)methanol

OR9781-1G OR9781-250MG
APOSOR9781-1GEA 6450 CZK
APOSOR9781-1G APOSOR9781-250MG
2-(Hydroxymethyl)-5-phenoxyfuran 97%
2-(Hydroxymethyl)-5-phenoxyfuran
Vzorec: C₁₁H₁₀O₃
MW: 190,195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08271913
CAS číslo: 51551-74-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR