2-Chloro-4-iodopyridin-3-ol 97%
Dodavatel: Apollo Scientific
OR968326-1G
OR968326-500MG
APOSOR968326-1GEA
5130
CZK
APOSOR968326-1G
APOSOR968326-500MG
2-Chloro-4-iodopyridin-3-ol 97%
2-Chloro-4-iodopyridin-3-ol
Vzorec:
C₅H₃ClINO Storage Temperature: Refrigerator |
MDL Number:
MFCD16610456 CAS číslo: 1207973-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...