Tisk…

N-Methyl-3-(thien-3-yl)benzylamine

Dodavatel: Apollo Scientific

Synonyms: 3-{[3-(Methylamino)methyl]phenyl}thiophene, N-Methyl-N-(3-thien-3-ylbenzyl)amine

OR9664-1G OR9664-250MG
APOSOR9664-1GEA 10350 CZK
APOSOR9664-1G APOSOR9664-250MG
N-Methyl-3-(thien-3-yl)benzylamine
N-Methyl-3-(thien-3-yl)benzylamine
Vzorec: C₁₂H₁₃NS
MW: 203.303 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08435893
CAS číslo: 884507-27-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR