Tisk…

4-Amino-2-phenylpyrimidine 97%

Dodavatel: Apollo Scientific

Synonyms: 2-Phenylpyrimidin-4-amine

OR965812-1G OR965812-250MG
APOSOR965812-1GEA 5130 CZK
APOSOR965812-1G APOSOR965812-250MG
4-Amino-2-phenylpyrimidine 97%
4-Amino-2-phenylpyrimidine
Vzorec: C₁₀H₉N₃
Storage Temperature: Ambient
MDL Number: MFCD11043737
CAS číslo: 33630-25-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR