Tisk…

4,7-Diazaindole 95%

Dodavatel: Apollo Scientific

Synonyms: 5H-Pyrrolo[2,3-b]pyrazine

OR965780-100MG OR965780-1G OR965780-250MG
APOSOR965780-100MGEA 2360 CZK
APOSOR965780-100MG APOSOR965780-1G APOSOR965780-250MG
4,7-Diazaindole 95%
4,7-Diazaindole
Vzorec: C₆H₅N₃
MW: 119,124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834819
CAS číslo: 42430-74-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR