Tisk…

1-(Ethoxycarbonyl)-3-cyclopentene-1-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR965675-1G OR965675-250MG
APOSOR965675-1GEA 15310 CZK
APOSOR965675-1G APOSOR965675-250MG
1-(Ethoxycarbonyl)-3-cyclopentene-1-carboxylic acid 95%
1-(Ethoxycarbonyl)-3-cyclopentene-1-carboxylic acid
Vzorec: C₉H₁₂O₄
MW: 184,189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115568
CAS číslo: 76910-08-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR