2-(4-Isobutylphenyl)acetic acid 95%
Dodavatel: Apollo Scientific
Synonyms:
Ibufenac
OR965605-1G
APOSOR965605-1GEA
1820
CZK
APOSOR965605-1G
2-(4-Isobutylphenyl)acetic acid 95%
2-(4-Isobutylphenyl)acetic acid
Vzorec:
C₁₂H₁₆O₂ MW: 192,254 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00864374 CAS číslo: 1553-60-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...