Tisk…

3-(Isothiocyanatomethyl)pyridine 95%

Dodavatel: Apollo Scientific

OR965604-1G OR965604-250MG
APOSOR965604-1GEA 16340 CZK
APOSOR965604-1G APOSOR965604-250MG
3-(Isothiocyanatomethyl)pyridine 95%
3-(Isothiocyanatomethyl)pyridine
Vzorec: C₇H₆N₂S
Storage Temperature: Ambient
MDL Number: MFCD00175166
CAS číslo: 36810-90-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR