N-[4-(Dimethylamino)benzyl]-N-ethylamine 95%
Dodavatel: Apollo Scientific
OR965584-1G
OR965584-250MG
OR965584-5G
APOSOR965584-1GEA
6850
CZK
APOSOR965584-1G
APOSOR965584-250MG
APOSOR965584-5G
N-[4-(Dimethylamino)benzyl]-N-ethylamine 95%
N-[4-(Dimethylamino)benzyl]-N-ethylamine
Vzorec:
C₁₁H₁₈N₂ MW: 178,274 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00465457 CAS číslo: 313552-99-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...