Tisk…

3-(Cyclopropylmethoxy)propan-1-amine 98%

Dodavatel: Apollo Scientific

OR965559-1G OR965559-250MG
APOSOR965559-1GEA 17990 CZK
APOSOR965559-1G APOSOR965559-250MG
3-(Cyclopropylmethoxy)propan-1-amine 98%
3-(Cyclopropylmethoxy)propan-1-amine
Vzorec: C₇H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD07329912
CAS číslo: 2986-60-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR