Tisk…

1-(3,4-Dihydro-2H-chromen-3-yl)methanamine hydrochloride 97%

Dodavatel: Apollo Scientific

Synonyms: Chroman-3-ylmethanamine hydrochloride

OR965546-1G OR965546-250MG
APOSOR965546-1GEA 11380 CZK
APOSOR965546-1G APOSOR965546-250MG
1-(3,4-Dihydro-2H-chromen-3-yl)methanamine hydrochloride 97%
1-(3,4-Dihydro-2H-chromen-3-yl)methanamine hydrochloride
Vzorec: C₁₀H₁₃NO·ClH
MW: 199,677 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07330841
CAS číslo: 113771-75-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR