Tisk…

N-[2-(Aminomethyl)benzyl]-n,n-dimethylamine 98%

Dodavatel: Apollo Scientific

OR965536-1G OR965536-5G
APOSOR965536-1GEA 5790 CZK
APOSOR965536-1G APOSOR965536-5G
N-[2-(Aminomethyl)benzyl]-n,n-dimethylamine 98%
N-[2-(Aminomethyl)benzyl]-n,n-dimethylamine
Vzorec: C₁₀H₁₆N₂
MW: 164,247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07324744
CAS číslo: 53369-77-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR