Tisk…

4-(2-Methoxyethoxy)benzenesulphonyl chloride 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-(2-Methoxyethoxy)-phenylsulphonyl chloride

OR965534-1G OR965534-250MG
APOSOR965534-1GEA 7740 CZK
APOSOR965534-1G APOSOR965534-250MG
4-(2-Methoxyethoxy)benzenesulphonyl chloride 95%
4-(2-Methoxyethoxy)benzenesulphonyl chloride
Vzorec: C₉H₁₁ClO₄S
MW: 250,699 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09816161
CAS číslo: 204072-53-7
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR