5-Aminomethyl-1H-pyridin-2-one 98%
Dodavatel: Apollo Scientific
Synonyms:
5-(Aminomethyl)pyridin-2-ol
OR965527-1G
OR965527-250MG
OR965527-5G
APOSOR965527-1GEA
7540
CZK
APOSOR965527-1G
APOSOR965527-250MG
APOSOR965527-5G
5-Aminomethyl-1H-pyridin-2-one 98%
5-Aminomethyl-1H-pyridin-2-one
Vzorec:
C₆H₈N₂O MW: 124,141 g/mol Bod tání: 170…200 °C Storage Temperature: Ambient |
MDL Number:
MFCD08274990 CAS číslo: 131052-84-1 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...