3-Phenyl-pent-1-yn-3-ol 95%
Dodavatel: Apollo Scientific
OR965295-1G
OR965295-250MG
APOSOR965295-1GEA
37690
CZK
APOSOR965295-1G
APOSOR965295-250MG
3-Phenyl-pent-1-yn-3-ol 95%
3-Phenyl-pent-1-yn-3-ol
Vzorec:
C₁₁H₁₂O Storage Temperature: Ambient |
MDL Number:
MFCD17926462 CAS číslo: 6712-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...