3-(3,4-Dihydro-4-methyl-3-oxoquinoxalin-2-yl)propanoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR9652-1G
OR9652-250MG
APOSOR9652-1GEA
4570
CZK
APOSOR9652-1G
APOSOR9652-250MG
3-(3,4-Dihydro-4-methyl-3-oxoquinoxalin-2-yl)propanoic acid
3-(3,4-Dihydro-4-methyl-3-oxoquinoxalin-2-yl)propanoic acid
Vzorec:
C₁₂H₁₂N₂O₃ Bod tání: 180…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00665740 CAS číslo: 1501-38-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...