1-(2-Methoxy-phenyl)-n1,n1-dimethyl-ethane-1,2-diamine 98%
Dodavatel: Apollo Scientific
OR965106-1G
OR965106-250MG
APOSOR965106-1GEA
17990
CZK
APOSOR965106-1G
APOSOR965106-250MG
1-(2-Methoxy-phenyl)-n1,n1-dimethyl-ethane-1,2-diamine 98%
1-(2-Methoxy-phenyl)-n1,n1-dimethyl-ethane-1,2-diamine
Vzorec:
C₁₁H₁₈N₂O MW: 194,273 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06655050 CAS číslo: 842976-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...