Cyclopropyl-thiophen-2-ylmethyl-amine 98%
Dodavatel: Apollo Scientific
OR965054-1G
OR965054-5G
APOSOR965054-1GEA
8270
CZK
APOSOR965054-1G
APOSOR965054-5G
Cyclopropyl-thiophen-2-ylmethyl-amine 98%
Cyclopropyl-thiophen-2-ylmethyl-amine
Vzorec:
C₈H₁₁NS Storage Temperature: Ambient |
MDL Number:
MFCD03856576 CAS číslo: 14471-18-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...