Tisk…

3-Chloro-N-phenylpropanamide 97%

Dodavatel: Apollo Scientific

OR964996-1G OR964996-5G
APOSOR964996-1GEA 2240 CZK
APOSOR964996-1G APOSOR964996-5G
3-Chloro-N-phenylpropanamide 97%
3-Chloro-N-phenylpropanamide
Vzorec: C₉H₁₀ClNO
MW: 183,635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00018971
CAS číslo: 3460-04-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR