3-Bromophthalic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR964924-5G
APOSOR964924-5GEA
2980
CZK
APOSOR964924-5G
3-Bromophthalic acid 98%
3-Bromophthalic acid
Vzorec:
C₈H₅BrO₄ MW: 245,027 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17069368 CAS číslo: 116-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...