Tisk…
N-(3-Aminopropyl)-acetamide hydrochloride 97%
Dodavatel: Apollo Scientific
OR964919-1G OR964919-500MG
APOSOR964919-1GEA 11180 CZK
APOSOR964919-1G APOSOR964919-500MG
N-(3-Aminopropyl)-acetamide hydrochloride 97%
N-(3-Aminopropyl)-acetamide hydrochloride
Vzorec: C₅H₁₂N₂O·ClH
MW: 152,623 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13196104
CAS číslo: 4078-13-1