Tisk…

Methyl 2,3-dihydrobenzofuran-5-carboxylate 95%

Dodavatel: Apollo Scientific

OR964912-1G OR964912-5G
APOSOR964912-1GEA 870 CZK
APOSOR964912-1G APOSOR964912-5G
Methyl 2,3-dihydrobenzofuran-5-carboxylate 95%
Methyl 2,3-dihydrobenzofuran-5-carboxylate
Vzorec: C₁₀H₁₀O₃
MW: 178,185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00114411
CAS číslo: 103204-80-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR