Tisk…

2-Amino-4-(chloromethyl)pyridine hydrochloride 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Amino-4-(chloromethyl)pyridine Hydrochloride

OR964731-1G
APOSOR964731-1GEA 3810 CZK
APOSOR964731-1G
2-Amino-4-(chloromethyl)pyridine hydrochloride 95%
2-Amino-4-(chloromethyl)pyridine hydrochloride
Vzorec: C₆H₇ClN₂·ClH
MW: 179,047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12198864
CAS číslo: 1186663-31-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR