Tisk…

4-Ethoxy-2-hydroxy-benzaldehyde 95%

Dodavatel: Apollo Scientific

Synonyms: 4-Ethoxy-2-hydroxybenzaldehyde

OR964660-1G OR964660-250MG
APOSOR964660-1GEA 2900 CZK
APOSOR964660-1G APOSOR964660-250MG
4-Ethoxy-2-hydroxy-benzaldehyde 95%
4-Ethoxy-2-hydroxy-benzaldehyde
Vzorec: C₉H₁₀O₃
MW: 166,174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03032734
CAS číslo: 43057-77-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR