Azepane-1-sulphonyl chloride 98%
Dodavatel: Apollo Scientific
OR964622-1G
OR964622-5G
APOSOR964622-1GEA
7670
CZK
APOSOR964622-1G
APOSOR964622-5G
Azepane-1-sulphonyl chloride 98%
Azepane-1-sulphonyl chloride
Vzorec:
C₆H₁₂ClNO₂S MW: 197.683 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08444495 CAS číslo: 41483-72-1 |