Tisk…

Methyl 3-oxo-3-(pyridin-4-yl)propanoate 95%

Dodavatel: Apollo Scientific

OR964608-1G OR964608-250MG OR964608-5G
APOSOR964608-1GEA 3770 CZK
APOSOR964608-1G APOSOR964608-250MG APOSOR964608-5G
Methyl 3-oxo-3-(pyridin-4-yl)propanoate 95%
Methyl 3-oxo-3-(pyridin-4-yl)propanoate
Vzorec: C₉H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD00466380
CAS číslo: 829-45-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR