Tisk…

2-Chloro-4-methylthiazole 96%

Dodavatel: Apollo Scientific

OR964534-10G OR964534-1G OR964534-5G
APOSOR964534-10GEA 4300 CZK
APOSOR964534-10G APOSOR964534-1G APOSOR964534-5G
2-Chloro-4-methylthiazole 96%
2-Chloro-4-methylthiazole
Vzorec: C₄H₄ClNS
Storage Temperature: Ambient
MDL Number: MFCD00051033
CAS číslo: 26847-01-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR