3-Amino-1-N-alloc-piperidine 95%
Dodavatel: Apollo Scientific
OR964532-1G
OR964532-250MG
APOSOR964532-1GEA
18420
CZK
APOSOR964532-1G
APOSOR964532-250MG
3-Amino-1-N-alloc-piperidine 95%
3-Amino-1-N-alloc-piperidine
Vzorec:
C₉H₁₆N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD03839997 CAS číslo: 886363-44-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...