4-(Pyridin-2-yloxy)benzoyl chloride
Dodavatel: Apollo Scientific
OR9644-1G
OR9644-250MG
APOSOR9644-1GEA
14520
CZK
APOSOR9644-1G
APOSOR9644-250MG
4-(Pyridin-2-yloxy)benzoyl chloride
4-(Pyridin-2-yloxy)benzoyl chloride
Vzorec:
C₁₂H₈ClNO₂ MW: 233.65 g/mol Bod tání: 79.5…80.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD08435898 CAS číslo: 51363-01-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...