Tisk…

3-Phenoxybenzoyl chloride 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR9642-1G OR9642-250MG
APOSOR9642-1GEA 5360 CZK
APOSOR9642-1G APOSOR9642-250MG
3-Phenoxybenzoyl chloride 97%
3-Phenoxybenzoyl chloride
Vzorec: C₁₃H₉ClO₂
Bod varu: 174…175 °C (12 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03424712
CAS číslo: 3586-15-0
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR