Tisk…

tert-Butyl thiazolidine-3-carboxylate 95%

Dodavatel: Apollo Scientific

OR964172-1G OR964172-250MG
APOSOR964172-1GEA 2780 CZK
APOSOR964172-1G APOSOR964172-250MG
tert-Butyl thiazolidine-3-carboxylate 95%
tert-Butyl thiazolidine-3-carboxylate
Vzorec: C₈H₁₅NO₂S
MW: 189,275 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22581582
CAS číslo: 148312-55-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR