2-Methylthieno[3,2-c]pyridin-4(5H)-one 95%
Dodavatel: Apollo Scientific
OR964039-1G
OR964039-250MG
APOSOR964039-1GEA
10220
CZK
APOSOR964039-1G
APOSOR964039-250MG
2-Methylthieno[3,2-c]pyridin-4(5H)-one 95%
2-Methylthieno[3,2-c]pyridin-4(5H)-one
Vzorec:
C₈H₇NOS MW: 165,212 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11215545 CAS číslo: 59207-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...