2-Methyl-3-n,n-dimethylaminomethylindole 95%
Dodavatel: Apollo Scientific
OR964014-1G
OR964014-250MG
APOSOR964014-1GEA
9720
CZK
APOSOR964014-1G
APOSOR964014-250MG
2-Methyl-3-n,n-dimethylaminomethylindole 95%
2-Methyl-3-n,n-dimethylaminomethylindole
Vzorec:
C₁₂H₁₆N₂ Storage Temperature: Ambient |
MDL Number:
MFCD00022712 CAS číslo: 37125-92-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...