2-Ethyl-2-methyl-chroman-4-one 95%
Dodavatel: Apollo Scientific
OR963935-1G
OR963935-250MG
APOSOR963935-1GEA
22950
CZK
APOSOR963935-1G
APOSOR963935-250MG
2-Ethyl-2-methyl-chroman-4-one 95%
2-Ethyl-2-methyl-chroman-4-one
Vzorec:
C₁₂H₁₄O₂ MW: 190,238 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07369953 CAS číslo: 73509-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...