Tisk…

2-Cyclohexylbenzaldehyde 95%

Dodavatel: Apollo Scientific

OR963923-1G OR963923-250MG
APOSOR963923-1GEA 8340 CZK
APOSOR963923-1G APOSOR963923-250MG
2-Cyclohexylbenzaldehyde 95%
2-Cyclohexylbenzaldehyde
Vzorec: C₁₃H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD06658151
CAS číslo: 128323-04-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR