Tisk…

2-Cyano-3-phenylpropionic acid 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Cyano-3-phenylpropionic Acid

OR963908-1G OR963908-250MG
APOSOR963908-1GEA 4300 CZK
APOSOR963908-1G APOSOR963908-250MG
2-Cyano-3-phenylpropionic acid 95%
2-Cyano-3-phenylpropionic acid
Vzorec: C₁₀H₉NO₂
MW: 175,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00205073
CAS číslo: 5331-42-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR