Tisk…

H-Pro-OEt hydrochloride 97%

Dodavatel: Apollo Scientific

Synonyms: (S)-Ethyl pyrrolidine-2-carboxylate hydrochloride

OR963811-25G
APOSOR963811-25GEA 3540 CZK
APOSOR963811-25G
H-Pro-OEt hydrochloride 97%
H-Pro-OEt hydrochloride
Vzorec: C₇H₁₃NO₂·ClH
MW: 179,645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191004
CAS číslo: 33305-75-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR