(S)-Methyl 2-amino-2-cyclopropylacetate hydrochloride 95%
Dodavatel: Apollo Scientific
OR963730-1G
OR963730-250MG
APOSOR963730-1GEA
3060
CZK
APOSOR963730-1G
APOSOR963730-250MG
(S)-Methyl 2-amino-2-cyclopropylacetate hydrochloride 95%
(S)-Methyl 2-amino-2-cyclopropylacetate hydrochloride
Vzorec:
C₆H₁₁NO₂·ClH MW: 165,618 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09607984 CAS číslo: 138326-68-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...