Tisk…

2-(6-Chloropyrazin-2-ylamino)acetic acid 95%

Dodavatel: Apollo Scientific

OR963015-1G OR963015-250MG
APOSOR963015-1GEA 16340 CZK
APOSOR963015-1G APOSOR963015-250MG
2-(6-Chloropyrazin-2-ylamino)acetic acid 95%
2-(6-Chloropyrazin-2-ylamino)acetic acid
Vzorec: C₆H₆ClN₃O₂
MW: 187,584 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12068353
CAS číslo: 1159822-45-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR