1-(5-Methyl-1H-pyrrol-3-yl)ethanone 95%
Dodavatel: Apollo Scientific
OR962933-1G
OR962933-250MG
APOSOR962933-1GEA
9720
CZK
APOSOR962933-1G
APOSOR962933-250MG
1-(5-Methyl-1H-pyrrol-3-yl)ethanone 95%
1-(5-Methyl-1H-pyrrol-3-yl)ethanone
Vzorec:
C₇H₉NO MW: 123.152 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19219386 CAS číslo: 6115-72-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...