Tisk…

2-(2-Chlorophenyl)propan-2-ol 95%

Dodavatel: Apollo Scientific

OR962887-1G OR962887-5G
APOSOR962887-1GEA 3770 CZK
APOSOR962887-1G APOSOR962887-5G
2-(2-Chlorophenyl)propan-2-ol 95%
2-(2-Chlorophenyl)propan-2-ol
Vzorec: C₉H₁₁ClO
MW: 170,636 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11870103
CAS číslo: 3670-15-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR