Tisk…

(4-Bromothiophen-2-yl)methanamine hydrobromide 95%

Dodavatel: Apollo Scientific
Varování

OR962877-1G OR962877-250MG
APOSOR962877-1GEA 9330 CZK
APOSOR962877-1G APOSOR962877-250MG
(4-Bromothiophen-2-yl)methanamine hydrobromide 95%
(4-Bromothiophen-2-yl)methanamine hydrobromide
Vzorec: C₅H₆BrNS·BrH
MW: 272,989 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11855809
CAS číslo: 1314707-83-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR