2-(2-Azabicyclo[2.2.1]heptan-2-yl)acetic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR962876-1G
OR962876-250MG
APOSOR962876-1GEA
0
CZK
APOSOR962876-1G
APOSOR962876-250MG
2-(2-Azabicyclo[2.2.1]heptan-2-yl)acetic acid 97%
2-(2-Azabicyclo[2.2.1]heptan-2-yl)acetic acid
Vzorec:
C₈H₁₃NO₂ MW: 155,194 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09055404 CAS číslo: 933690-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...