tert-Butyl N-(3-iodopropyl)carbamate 96%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
tert-Butyl (3-iodopropyl)carbamate
OR962795-1G
OR962795-5G
APOSOR962795-1GEA
4400
CZK
APOSOR962795-1G
APOSOR962795-5G
tert-Butyl N-(3-iodopropyl)carbamate 96%
tert-Butyl N-(3-iodopropyl)carbamate
Vzorec:
C₈H₁₆INO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD09951819 CAS číslo: 167479-01-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...